Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nerol, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
106-25-2
C10H18O
154.25
MFCD00063204
GLZPCOQZEFWAFX-YFHOEESVSA-N
nerol,cis-geraniol,neryl alcohol,z-3,7-dimethylocta-2,6-dien-1-ol,z-geraniol,2z-3,7-dimethylocta-2,6-dien-1-ol,2,6-octadien-1-ol, 3,7-dimethyl-, 2z,nerol natural,cis-3,7-dimethyl-2,6-octadien-1-ol,z-3,7-dimethyl-2,6-octadien-1-ol
643820
CHEBI:29452
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
CC(=CCCC(=CCO)C)C
106-25-2
C10H18O
154.25
MFCD00063204
GLZPCOQZEFWAFX-YFHOEESVSA-N
nerol,cis-geraniol,neryl alcohol,z-3,7-dimethylocta-2,6-dien-1-ol,z-geraniol,2z-3,7-dimethylocta-2,6-dien-1-ol,2,6-octadien-1-ol, 3,7-dimethyl-, 2z,nerol natural,cis-3,7-dimethyl-2,6-octadien-1-ol,z-3,7-dimethyl-2,6-octadien-1-ol
643820
CHEBI:29452
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
CC(=CCCC(=CCO)C)C
Flash Point 100°C
Boiling Point 225°C to 227°C
Specific Gravity 0.875
Refractive Index 1.4720 to 1.479
Packaging Glass bottle
Quantity 500mL
Infrared Spectrum Authentic
Density 0.8750g/mL
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2CCH(CH2)2C(CH3)CHCH2OH
Beilstein 01, IV, 2276
Merck Index 15, 6560
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in absolute alcohol
Formula Weight 154.25
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Nerol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-378-7