Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neryl acetate, mixture of ca 55-65% neryl and 35-45% geranyl acetate, 97%, Thermo Scientific™

Catalog No. AC296230250
Click to view available options
Quantity:
25 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-12-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00063205
InChI Key HIGQPQRQIQDZMP-FLIBITNWSA-N
Synonym neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h
PubChem CID 1549025
IUPAC Name [(2Z)-3,7-dimethylocta-2,6-dienyl] acetate
SMILES CC(=CCCC(=CCOC(=O)C)C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.91g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4590 to 1.4620 (20°C, 589nm)
Linear Formula CH3CO2CH2CHC(CH3)CH2CH2CHC(CH3)2
Quantity 25 g
Formula Weight 196.29
Percent Purity ≥96.0% (sum of isomers)
Physical Form Liquid
Chemical Name or Material Neryl acetate, mixture of ca 55-65% neryl and 35-45% geranyl acetate, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.