Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nesfatin-1/Nucleobindin-2 Antibody (2062A), Alexa Fluor 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB59491AF532
Description
Nesfatin-1/Nucleobindin-2 Monoclonal specifically detects Nesfatin-1/Nucleobindin-2 in Human samples. It is validated for Western Blot.Specifications
Nesfatin-1/Nucleobindin-2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NUCB2 | |
Partial recombinant human NUCB2/Nesfatin-1 protein (amino acids 25-106) [UniProt P80303] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
2062A | |
Western Blot | |
DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, nucleobindin 2, nucleobindin-2, nucleobinding 2 | |
Rabbit | |
Protein A or G purified | |
Signal Transduction | |
4925.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction