Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nesprin 1 Monoclonal Antibody (OTI1E10)
GREENER_CHOICE

Catalog No. PIMA527519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527519 Supplier Invitrogen™ Supplier No. MA527519
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nesprin 1 Monoclonal Antibody for Western Blot, IHC (P)

There are two genes encoding members of a new family of type II integral membrane proteins. Both are ubiquitously expressed, and tissue-specific alternative mRNA initiation and splicing generate at least two major isoforms of each protein, with the smaller isoforms being truncated at the N-terminus. These proteins are called nesprin-l and -2 for nuclear _envelope _spectrin repeat, as they are characterized by the presence of multiple, clustered spectrin repeats, bipartite nuclear localization sequences and a conserved C-terminal, single transmembrane domain. Transient transfection of EGFP-fusion expression constructs demonstrated their localization to the nuclear membrane with a novel C-terminal, TM-domain-containing sequence essential for perinuclear localization. Nesprin-l is developmentally regulated in both smooth and skeletal muscle and is relocalized from the nuclear envelope to the nucleus and cytoplasm during C2Cl2 myoblast differentiation. Nesprins may function as 'dystrophins of the nucleus' to maintain nuclear organization and structura integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nesprin 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYNE1
Gene Accession No. Q8NF91
Gene Alias 8B; A330049M09Rik; ARCA1; BE692247; C130039F11Rik; C6orf98; CPG2; CPG2 full length; dJ45H2.2; DKFZp781J13156; EDMD4; enaptin; enaptin165; FLJ30878; FLJ41140; KASH domain-containing protein 1; KASH1; KIAA0796; KIAA1262; KIAA1756; mKIAA1756; MYNE1; Myne-1; Myocyte nuclear envelope protein 1; Nesp1; nesprin 1; nesprin-1; Nuclear envelope spectrin repeat protein 1; RP1-130E4.2; SCAR8; spectrin repeat containing nuclear envelope protein 1; spectrin repeat containing, nuclear envelope 1; synaptic nuclear envelope 1; synaptic nuclear envelope protein 1; synaptic nuclei expressed gene 1; SYNE 1; SYNE1; Syne-1; syne-1 fragment
Gene Symbols SYNE1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-285 of SYNE1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23345
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.