Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nesprin 2 Antibody (10H8), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP259945F

Catalog No. NBP2-59945F


Only null left
Add to Cart

Description

Description

Nesprin 2 Monoclonal specifically detects Nesprin 2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nesprin 2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
SYNE2
GST fused to residues 6,653-6,835 of human Nesprin 2
0.1 mL
Primary
Specificity of human Nesprin 2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C in the dark.
Immunocytochemistry, Immunofluorescence
10H8
Immunocytochemistry/Immunofluorescence
DKFZP434H2235, DKFZp686E01115, DKFZp686H1931, EDMD5, FLJ46790, KIAA1011FLJ11014, nesprin 2, nesprin-2, NUAFLJ43727, NUANCEFLJ45710, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, polytrophin, Protein NUANCE, spectrin repeat containing, nuclear envelope 2, Synaptic nuclear envelope protein 2, synaptic nuclei expressed gene 2, SYNE-2, TROPH
Mouse
Protein A purified
Developmental Biology
23224
Human
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.