Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nesprin-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24917525UL
Description
Nesprin-3 Polyclonal antibody specifically detects Nesprin-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Nesprin-3 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
chromosome 14 open reading frame 49, FLJ25605, MGC75267, nesprin-3, NET53 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MKAEYDAVKAKAQKRVDLLEQVAREHEEYQAGVDEFQLWLKAVVEKVNGCLGRNCKLPITQRLSTLQDIAKDFPRGEESLETLEEQSAGVIRNTSP | |
25 μL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
161176 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction