Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NESS 0327
GREENER_CHOICESDP

Catalog No. 574650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-465-0 50 mg
57-461-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-465-0 Supplier Tocris Bioscience™ Supplier No. 5746/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra-high affinity and selective CB1 antagonist

NESS 0327 is an ultra-high affinity and selective CB1 receptor antagonist (Ki = 350 fM). Exhibits >60-fold selectivity for CB1 over CB2 receptors. Antagonizes the inhibitory effects of WIN 55,212-2 (Cat. No. 1038) on electrically evoked contractions in mouse isolated vas deferens ex vivo. Also antagonizes the antinociceptive effect of WIN 55,212-2 in tail-flick and hot plate tests in vivo.

Specifications

Chemical Name or Material 8-Chloro-1-(2,4-dichlorophenyl)-1,4,5,6-tetrahydro-N-1-piperidinyl-benzo[6,7 ]cyclohepta[1,2-c]pyrazole-3-carboxamide
CAS 494844-07-4
Quantity 50 mg
Target Cannabinoid R1/CB1 Antagonists
Molecular Formula C24H23Cl3N4O
Formula Weight 7676
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.