Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nessler Reagent, for Ammonia Nitrogen Determination, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Volume (Metric):
120 mL
500 mL
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
Description
- For Ammonia Nitrogen Determination
Specifications
Specifications
| Color | Clear light yellow |
| Volume (Metric) | 1 L |
| Format | Liquid |
| Packaging | Amber Poly Bottle |
| Quantity | 1 L |
| Sterility | Non-sterile |
| Type | Nessler Reagent |
| Volume (English) | 0.3 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction