Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nessler's Reagent (Mercuric - Potassium Iodide TS, Alkaline), USP 25 Formulation, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Color | Clear light yellow |
| pH Range | >12 |
| Volume (Metric) | 1 L |
| Format | Liquid |
| Packaging | Amber Poly Bottle |
| Quantity | 1 L |
| Type | Nessler's Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction