Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Nestable Bucket Balance
SCI_ED

Catalog No. S01292
Click to view available options
Class Size:
1 Student
4 Students

Save space and encourage children to explore measurement and weight. Both balances feature an efficient, compact design with equivalency markings that allow children to assign less than, greater than, or equal to symbols to their balance setup and demonstrate mass equivalency

  • The math balance encourages kids to explore measurement and weight.
  • Place math manipulatives, counting bears, or math counters in the balancing scale to learn about their weight difference.
  • Students will learn early mathematical and scientific concepts such as inequalities, mass, volume, operations, counting, subtracting, and adding.
  • The durable clear plastic bucket balance allows children to explore volume and compare solids and liquids. Kids get to develop observation and estimation skills while exploring volume and measurements.
  • The equivalency markings allow students to assign less than, greater than, or equal to symbols to their balance setup.
  • Teachers, parents, or kids can easily demonstrate mass equivalency.
  • Made from high quality plastic, the school balance bucket can be assembled and disassembled in 3 quick and easy steps.
  • The Nestable Bucket Balance features an efficient, compact design that reduces storage space by 80%.
  • Perfect for the classroom, daycare, or homeschool.
  • Students in activity-based programs perform up to 20% higher than groups using traditional or textbook approaches.
  • For over 50 years, hand2mind has encouraged hands-on learning and discovery, driving deeper understanding and helping children unlock their full potential. With products designed to enhance learning in the classroom and the world around us, we strive to find new ways to encourage hands-on exploration and discovery.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Student
Product Type Balance
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.