Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nestin Antibody (10C2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300266MFV450
Description
Nestin Monoclonal antibody specifically detects Nestin in Human, Mouse, Rabbit (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Nestin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rabbit (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
10C2 | |
Flow Cytometry | |
FLJ21841, nestin | |
A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681] | |
0.1 mL | |
Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
10763 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction