Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nestin Antibody (4D11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192717AF594
Description
Nestin Monoclonal antibody specifically detects Nestin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
Nestin | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
4D11 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FLJ21841, nestin | |
Recombinant construct, amino acids 317-630 of the human Nestin protein expressed in and purified from E. coli. [UniProt P48681] | |
0.1 mL | |
Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
10763 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction