Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody (4D11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 17 publications

$236.98 - $618.54

Specifications

Antigen Nestin
Clone 4D11
Concentration 1 mg/ml
Dilution Western Blot 1:5000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP192717SS
SDP
View Documents
Novus Biologicals
NBP192717SS
0.025 mL
Each for $236.98
Only null left
Add to Cart
 
NBP192717
SDP
View Documents
Novus Biologicals
NBP192717
0.1 mL
Each for $618.54
Only null left
Add to Cart
 
Description

Description

Nestin Monoclonal specifically detects Nestin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nestin
1 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
50% PBS, 50% glycerol with 5mM Sodium Azide
FLJ21841, nestin
NES
IgG1
Protein G purified
240 kDa
4D11
Western Blot 1:5000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500
Monoclonal
Ascites
RUO
Human, Mouse, Rat
P48681
10763
Recombinant construct, amino acids 317-630 of the human Nestin protein expressed in and purified from E. coli. [UniProt P48681]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.