Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nestin Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300265AF647
Description
Nestin Polyclonal specifically detects Nestin in Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Product Image.Specifications
Nestin | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
P48681 | |
NES | |
Recombinant human Nestin protein (corresponding to residues 1464-1614). [UniProt# P48681] | |
0.1 mL | |
Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
10763 | |
Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLJ21841, nestin | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for human nestin. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction