Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody, Novus Biologicals™
SDP

Chicken Polyclonal Antibody has been used in 46 publications

$587.00

Specifications

Antigen Nestin
Classification Polyclonal
Conjugate Unconjugated
Host Species Chicken
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001604
SDP
View Documents
Novus Biologicals
NB1001604
0.25 mL
Each for $587.00
Only null left
Add to Cart
 
NB10016041M
SDP
View Documents
Novus Biologicals
NB10016041ML
1 mL N/A N/A N/A
Description

Description

Nestin Polyclonal specifically detects Nestin in Human, Mouse, Rat, Baboon, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Nestin
Unconjugated
RUO
FLJ21841, nestin
NES
Primary
Nestin
Polyclonal
Chicken
Stem Cell Markers
10763
IgY
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.