Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nestin Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300265PCP
Description
Nestin Polyclonal antibody specifically detects Nestin in Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Nestin | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
FLJ21841, nestin | |
Recombinant human Nestin protein (corresponding to residues 1464-1614). [UniProt# P48681] | |
0.1 mL | |
Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
10763 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction