Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Monoclonal Antibody (196908), APC

Nestin Monoclonal Antibody (196908), APC

Supplier:  Thermo Scientific MA523650

Catalog No. PIMA523650


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nestin
Monoclonal
APC
Nes
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin
Mouse
Protein A/G
RUO
10763
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
196908
Buffered saline solution with BSA and 0.09% sodium azide
P48681
Nes
NS0 mouse myeloma cell line transfected with human Nestin.
100 Tests
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.