Learn More
Invitrogen™ Nestin Monoclonal Antibody (3k1)

Description
Recommended positive controls: Human neural progenitors and A172 cells. MA1-26777 will not react with rodent. Keep as concentrated solution. Store at 4°C short term. For extended storage aliquot- add 50% glycerol and store at -20°C or below. Avoid freeze-thaw cycles.
Specifications
Specifications
| Antigen | Nestin |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3k1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and no preservative; pH 7.4 |
| Gene | Nes |
| Gene Accession No. | P48681, Q6P5H2 |
| Gene Alias | AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.