Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nestin Monoclonal Antibody (Rat-401 (4D4))
GREENER_CHOICE

Catalog No. MA181819
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA181819 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA181819 Supplier Invitrogen™ Supplier No. MA181819
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not react with human. In Western blot, this antibody detects a ~200-220 kDa band in newborn rat or mouse cell extracts. For immunohistochemistry, we recommend perfusing tissues with 4% paraformaldehyde (pH 7.4) for light microscopy and with either 4% paraformaldehyde (pH 10.0) or 4% paraformaldehyde with 0.1% glutaraldehyde (pH 7.4) for EM. For Immunocytochemistry we recommend using cells fixed in 4% paraformaldehyde buffered with 50mM sodium borate (pH 9.5). For Western blot, it is recommended that samples should be boiled in 4 volumes of 125 mM Tris, pH 6.8, 10% 2-mercaptoethanol, 10% glycerol and 4.6% SDS. Membranes should be blocked with milk or BSA. 5% PAGE gels are suggested. Mouse anti Rat Nestin antibody, clone Rat-401 recognizes rat nestin, a large intermediate filament protein transiently expressed in embryonic glian cells (Hockfield and McKay 1985).

Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Rat-401 (4D4)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene Nes
Gene Accession No. P21263, Q6P5H2
Gene Alias AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin
Gene Symbols Nes
Host Species Mouse
Immunogen Nestin purified from embryonic rat spinal cord.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18008, 25491
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.