Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NET1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10040800
Description
NET1 Polyclonal specifically detects NET1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
NET1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ARHGEF8neuroepithelial cell-transforming gene 1 protein, guanine nucleotide regulatory protein (oncogene), NET1A, neuroepithelial cell transforming 1, neuroepithelial cell transforming gene 1, neuroepithelioma transforming gene 1, p65 Net1 proto-oncogene protein, Proto-oncogene p65 Net1, Rho guanine nucleotide exchange factor (GEF) 8, Rho guanine nucleotide exchange factor 8, small GTP-binding protein regulator | |
Goat | |
Affinity Purified | |
RUO | |
10276 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry 2.5 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_005854.2 | |
NET1 | |
Peptide with sequence C-DKLEYLDEKQRDPR corresponding to internal region according to NP_001040625.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_001040625.1 and NP_005854.2). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction