Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NET1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102391 Supplier Invitrogen™ Supplier No. PA5102391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NET1.

Acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase. May be involved in activation of the SAPK/JNK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NET1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NET1
Gene Accession No. Q7Z628, Q9Z206
Gene Alias Arhgef8; guanine nucleotide regulatory protein (oncogene); mNET1; NET1; Net1 homolog; Net1a; neuroepithelial cell transforming 1; neuroepithelial cell transforming gene 1; neuroepithelial cell-transforming gene 1 protein; neuroepithelioma transforming gene 1; p65 Net1 proto-oncogene protein; Proto-oncogene p65 Net1; Rho guanine nucleotide exchange factor (GEF) 8; rho guanine nucleotide exchange factor 8; RP11-116G8.1; small GTP-binding protein regulator
Gene Symbols NET1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NET1(Accession Q7Z628), corresponding to amino acid residues A563-V596.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10276, 307098, 56349
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.