Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NET1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7096 Supplier Bioss Supplier No. BS6167R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase. May be involved in activation of the SAPK/JNK pathway.

Specifications

Antigen NET1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NET1
Gene Accession No. Q7Z628
Gene Alias ARHGEF8; guanine nucleotide regulatory protein (oncogene); mNET1; NET1; Net1 homolog; NET1A; neuroepithelial cell transforming 1; neuroepithelial cell transforming gene 1; Neuroepithelial cell-transforming gene 1 protein; neuroepithelioma transforming gene 1; p65 Net1 proto-oncogene protein; Proto-oncogene p65 Net1; Rho guanine nucleotide exchange factor (GEF) 8; Rho guanine nucleotide exchange factor 8; RP11-116G8.1; small GTP-binding protein regulator
Gene Symbols NET1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NET1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10276
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.