Learn More
Netilmicin sulfate, 595 ^mg/mg
Description
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
| Type | Netilmicin Sulfate |
| Physical Form | Powder |
| Color | White |
| Quantity | 10 mg |
| Sensitivity | Hygroscopic; Light sensitive |
| Boiling Point | 868°C |
| Merck Index | 14,6479 |
| Solubility Information | Soluble in water |
| Formula Weight | 720.75 |
| Chemical Name or Material | Netilmicin sulfate, 590μg/mg |
Safety and Handling
| Hazard Category | H360 |
| Hazard Statement | GHS H Statement H360-H312-H332 May damage fertility or the unborn child. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 260-147-3 |
| RTECSNumber | WK2286000 |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.