Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NETO1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241134
Description
NETO1 Polyclonal specifically detects NETO1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NETO1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NETO1 | |
Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human NETO1. The immunogen is located within amino acids 350 - 400 of NETO1. Amino Acid Squence: KQPRKKYVQRKSDFDQTV | |
Affinity Purified | |
RUO | |
Primary | |
NETO1 antibody is predicted to not cross-react with other NETO protein family members. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml | |
Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1 | |
Rabbit | |
60 kDa | |
0.1 mg | |
Vision | |
81832 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction