Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NETO1 Monoclonal Antibody (9A9E4)
GREENER_CHOICE

Catalog No. PIMA530701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530701 Supplier Invitrogen™ Supplier No. MA530701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NETO1.

This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. A similar gene in mice encodes a protein that plays a critical role in spatial learning and memory by regulating the function of synaptic N-methyl-D-aspartic acid receptor complexes in the hippocampus. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NETO1
Applications ELISA
Classification Monoclonal
Clone 9A9E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NETO1
Gene Accession No. Q8TDF5
Gene Alias AI851453; BCTL1; brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Btcl1; C130005O10Rik; NETO1; neuopilin- and tolloid-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin and tolloid like 1; neuropilin- and tolloid-like protein 1; neuropilin and tolloid-like protein 1; RGD1566269; si:dkey-82k12.12
Gene Symbols NETO1
Host Species Mouse
Immunogen Recombinant Human NETO1 Protein (Met1-Thr344).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.