Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NETO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566699 Supplier Invitrogen™ Supplier No. PA566699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PIIGRFCGQQN PPVIKSSGRF LWIKFFADGE LESMGFSARY NFTPDPDFKD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. A similar gene in mice encodes a protein that plays a critical role in spatial learning and memory by regulating the function of synaptic N-methyl-D-aspartic acid receptor complexes in the hippocampus. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NETO1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NETO1
Gene Accession No. Q8TDF5
Gene Alias AI851453; BCTL1; brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Btcl1; C130005O10Rik; NETO1; neuopilin- and tolloid-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin and tolloid like 1; neuropilin- and tolloid-like protein 1; neuropilin and tolloid-like protein 1; RGD1566269; si:dkey-82k12.12
Gene Symbols NETO1
Host Species Rabbit
Immunogen Recombinant Human NETO1. Recombinant protein control fragment (Product #RP-107350).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.