Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NETO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572701
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572701 Supplier Invitrogen™ Supplier No. PA572701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NETO1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. A similar gene in mice encodes a protein that plays a critical role in spatial learning and memory by regulating the function of synaptic N-methyl-D-aspartic acid receptor complexes in the hippocampus. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NETO1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NETO1
Gene Accession No. Q8R4I7, Q8TDF5
Gene Alias AI851453; BCTL1; brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Btcl1; C130005O10Rik; NETO1; neuopilin- and tolloid-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin and tolloid like 1; neuropilin- and tolloid-like protein 1; neuropilin and tolloid-like protein 1; RGD1566269; si:dkey-82k12.12
Gene Symbols NETO1
Host Species Rabbit
Immunogen NETO1 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human NETO1. The immunogen is within amino acids 350 - 400 of NETO1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246317, 307206, 81832
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.