Learn More
Invitrogen™ NETO2 Polyclonal Antibody

Description
Peptide sequence: ELSGADGIVR SSQVEQEEKT KPGQAVDCIW TIKATPKAKI YLRFLDYQME Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | NETO2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | NETO2 |
| Gene Accession No. | Q8NC67 |
| Gene Alias | 5530601C23Rik; AI854825; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2; BTCL2; FLJ10430; FLJ14724; FLJ90456; NEOT2; NEOT2FLJ14724; Neto2; neuropilin (NRP) and tolloid (TLL)-like 2; neuropilin and tolloid like 2; neuropilin and tolloid like-2; neuropilin and tolloid-like protein 2; UNQ1926/PRO4401 |
| Gene Symbols | NETO2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.