Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NETO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548040
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548040 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548040 Supplier Invitrogen™ Supplier No. PA548040
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human NETO1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

NETO2 is a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis.This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants have been observed, but they have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NETO2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NETO2
Gene Accession No. Q8BNJ6, Q8NC67
Gene Alias 5530601C23Rik; AI854825; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2; BTCL2; FLJ10430; FLJ14724; FLJ90456; NEOT2; NEOT2FLJ14724; Neto2; neuropilin (NRP) and tolloid (TLL)-like 2; neuropilin and tolloid like 2; neuropilin and tolloid like-2; neuropilin and tolloid-like protein 2; UNQ1926/PRO4401
Gene Symbols NETO2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human NETO2 isoform 1 Ile23-Lys345.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74513, 81831
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.