Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NETO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553029 Supplier Invitrogen™ Supplier No. PA553029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYPPNKECIY ILEAAPRQRI ELTFDEHYYI EPSFECRFDH LEVRDGPFGF SPLIDRYCGV KSPPLIRSTG RFMWIKFSSD EELEGLGFRA KYSFIPDPDF TYLGGILNPI PDCQFELSGA DGIVRSSQVE QEEKTKPGQA VDCIWTIKA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

NETO2 is a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis.This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants have been observed, but they have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NETO2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NETO2
Gene Accession No. Q8NC67
Gene Alias 5530601C23Rik; AI854825; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2; BTCL2; FLJ10430; FLJ14724; FLJ90456; NEOT2; NEOT2FLJ14724; Neto2; neuropilin (NRP) and tolloid (TLL)-like 2; neuropilin and tolloid like 2; neuropilin and tolloid like-2; neuropilin and tolloid-like protein 2; UNQ1926/PRO4401
Gene Symbols NETO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NETO2. Recombinant protein control fragment (Product #RP-90467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81831
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.