Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NETO2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981763
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1763 Supplier Bioss Supplier No. BS11896R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NETO2 is a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis.This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants have been observed, but they have not been fully characterized.

Specifications

Antigen NETO2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NETO2
Gene Accession No. Q8NC67
Gene Alias 5530601C23Rik; AI854825; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2; Btcl2; FLJ10430; FLJ14724; FLJ90456; LOC100349959; NEOT2; NEOT2FLJ14724; Neto2; neuropilin (NRP) and tolloid (TLL)-like 2; neuropilin and tolloid like 2; neuropilin and tolloid like-2; Neuropilin and tolloid-like protein 2; neuropilin- and tolloid-like protein 2-like; UNQ1926/PRO4401
Gene Symbols NETO2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NETO2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81831
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.