Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Netrin 4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987444
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7444 Supplier Bioss Supplier No. BS6711R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NTN4 contains 3 laminin EGF-like domains, 1 laminin N-terminal domain and 1 NTR domain. NTN4 may play an important role in neural, kidney and vascular development. It promotes neurite elongation from olfactory bulb explants. NTN4 belongs to a family of proteins related to laminins (see LAMA1, MIM 150320) Koch et al. (2000) [PubMed 11038171].[supplied by OMIM].

Specifications

Antigen Netrin 4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NTN4
Gene Accession No. Q9HB63
Gene Alias Beta-netrin; BOS_5266; CDY; CDY1A; FLJ23180; FLJ77185; Hepar-derived netrin-like protein; laminin B1-like; LOC100356958; netrin 4; Netrin-4; NTN4; PRO3091; RGD1565947
Gene Symbols NTN4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Netrin 4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59277
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.