Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEU-1/Sialidase-1 Antibody (OTI3D4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272942H
Description
NEU-1/Sialidase-1 Monoclonal specifically detects NEU-1/Sialidase-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NEU-1/Sialidase-1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NEU1 | |
Recombinant protein fragment corresponding to amino acids 48-315 of human NEU-1/Sialidase-1 (NP_000425)produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI3D4 | |
Western Blot, Immunohistochemistry | |
Acetylneuraminyl hydrolase, EC 3.2.1.18, exo-alpha-sialidase, FLJ93471, G9 sialidase, Lysosomal sialidase, N-acetyl-alpha-neuraminidase 1, NANH, NEU, SIAL1, sialidase 1 (lysosomal sialidase), sialidase-1 | |
Mouse | |
Affinity Purified | |
metabolism | |
4758.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction