Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554430 Supplier Invitrogen™ Supplier No. PA554430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDGVFCLLSD DHGASWRYGS GVSGIPYGQP KQENDFNPDE CQPYELPDGS VVINARNQNN YHCHCRIVLR SYDACDTLRP RDVTFDPELV DPVVAAGAVV TSSGIVFFSN PAHPEFRVNL TLRWSFSNG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

NEU1 is a lysosomal enzyme, which cleaves terminal sialic acid residues from substrates such as glycoproteins and glycolipids. In the lysosome, this enzyme is part of a heterotrimeric complex together with beta-galactosidase and cathepsin A. Mutations in NEU1 gene can lead to sialidosis.The protein encoded by this gene encodes the lysosomal enzyme, which cleaves terminal sialic acid residues from substrates such as glycoproteins and glycolipids. In the lysosome, this enzyme is part of a heterotrimeric complex together with beta-galactosidase and cathepsin A (the latter also referred to as 'protective protein'). Mutations in this gene can lead to sialidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEU1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEU1
Gene Accession No. Q99519
Gene Alias AA407268; AA407316; Acetylneuraminyl hydrolase; Aglp; Apl; Bat7; Bat-7; EC 3.2.1.18; exo-alpha-sialidase; FLJ93471; G9; G9 sialidase; HLA-B-associated transcript 7; lysosomal sialidase; Map-2; N-acetyl-alpha-neuraminidase 1; NANH; Neu; NEU1; Neu-1; neuraminidase 1; neuraminidase 1 (lysosomal sialidase); SIAL1; sialidase 1; sialidase 1 (lysosomal sialidase); sialidase-1
Gene Symbols NEU1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEU1. Recombinant protein control fragment (Product #RP-90541).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4758
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.