Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEU2 Monoclonal Antibody (OTI8C12), TrueMAB™, OriGene
SDP

Catalog No. 502540975
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0975 30 μL
50-167-4254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0975 Supplier OriGene Supplier No. TA503332S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEU2 Monoclonal antibody specifically detects NEU2 in Human samples. It is validated for Western Blot

Specifications

Antigen NEU2
Applications Western Blot
Classification Monoclonal
Clone OTI8C12
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NEK9
Gene Accession No. Q8TD19
Gene Alias Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; NEU2; neuraminidase 2; neuraminidase 2 (cytosolic sialidase); SIAL2; sialidase 2 (cytosolic sialidase); sialidase-2
Host Species Mouse
Immunogen Full length human recombinant protein of human NEK9 produced in HEK293 cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91754
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.