Learn More
NEU2 Mouse anti-Human, Clone: OTI8C12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503332

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.Specifications
NEU2 | |
Monoclonal | |
Unconjugated | |
NEU2 | |
SIAL2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8C12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y3R4 | |
NEU2 | |
Full length human recombit protein of human NEU2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.