Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neudesin/NENF Mouse anti-Human, Mouse, Clone: 4G9E12, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 601311IG150UL

Description
NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.Specifications
Neudesin/NENF | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CQ45, Q9UMX5 | |
Nenf | |
Neudesin/NENF Fusion Protein Ag8387 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4G9E12 | |
Unconjugated | |
NENF | |
CIR2, NENF, NEUDESIN, SCIRP10, SPUF, SPUF protein | |
Mouse | |
Protein A | |
RUO | |
29937, 66208 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction