Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuferricin Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309308100UL
Description
Neuferricin Polyclonal specifically detects Neuferricin in Rat samples. It is validated for Western Blot.Specifications
Neuferricin | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
cytochrome b5 domain containing 2, cytochrome b5 domain-containing protein 2, MGC32124, neuferricin | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Rat Neuferricin (NP_001007672). Peptide sequence EWSSAKGSRLWCSQKSGGVHRDWIGVPRKLYKPGAKEPHCVCVRTTGPPS | |
100 μg | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
124936 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction