Learn More
Eisco™ NeuLog Accessory Cable
SCI_ED
Catalog No. S9824350ND Shop All Eisco Products
Change view
Click to view available options
For Use With (Equipment):
NeuLog neuron logger sensors
Catalog No. For Use With (Equipment)
S98243-50ND NeuLog neuron logger sensors
1 options
This item is intended for educational purposes only.
Catalog No. S98243-50ND Supplier Eisco™ Supplier No. CBL261
This item is intended for educational purposes only.
Battery Module
  • Required to use sensors remotely or in the field
  • Powers sensors or chain of sensors operating off-line; connects sensors to RF Communication Module (S98243-46ND)
  • Rechargeable via PC USB port and data cable (sold separately)
  • Features LED charge indicator and push-button operation
Carrying Case
  • Constructed of black ballistic nylon
  • Holds up to 25 probes including cables and accessories
  • Measures 16 × 12 × 2.5 in. (40.6 × 30.5 × 6.4cm)
  • With shoulder strap
Specifications
Type Cable, Data
For Use With (Equipment) NeuLog neuron logger sensors
Length (Metric) 1 m