Learn More
Eisco™ Neulog Carbon Dioxide (CO2) Logger Sensor
SCI_ED
Catalog No. S139256 Shop All Eisco Products
Change view
Click to view available options
Response Time:
1 sec. to 31 days
Catalog No. Response Time
S139256 1 sec. to 31 days
1 options
This item is not returnable. View return policy
Catalog No. S139256 Supplier Eisco™ Supplier No. NUL260
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for collecting CO2 levels in the laboratory or in the field for up to 31 days and transmitting data to either your computer or mobile device for analysis with included Windows, IOS, or Android software.
  • The sensor comes pre-calibrated so you can start experimentation right out of the box.
  • Some fields of study include Chemistry, Physiology, Exercise Science, Biochemistry, Biology, Earth Science, Environmental Science, etc.
  • Among hundreds of possible experiments that can be performed with the NUL-260 sensor are: Monitoring ecological systems, combustion reactions, CO2 consumption rates, studying photosynthesis, testing chemical solutions, hydrogen peroxide decomposition, and many more.
  • Includes rugged plastic ergonomic case, attached CO2 probe, glass bottle for calibration, and instruction guide. Requires either USB-200 Module or WIFI-202 module, available separately.
  • Calibration: Factory-calibrated (offsets to standard atmospheric CO2 concentration of 380 ppm in open air). Must be calibrated before every use
Specifications
Type Sensor
Display Type Status Indicator Lights
Calibration Factory-calibrated
Alarm Types LED Indicator
Software Type Windows, IOS, Android
Measurement Range 350 to 10000 ppm
Resolution 1 ppm, ADC 14 bit
Response Time 1 sec. to 31 days