Learn More
Eisco™ NeuLog™ Wi-Fi Communication Module
SCI_ED
Catalog No. S23772 Shop All Eisco Products
Change view
Click to view available options
For Use With (Application):
Works with up to 5 sensors at once
Catalog No. For Use With (Application)
S23772 Works with up to 5 sensors at once
1 options
Catalog No. S23772 Supplier Eisco™ Supplier No. WIFI202
Add to Cart
Add to Cart
Add convenience and efficiency to your workflow. The NeuLog™ Wi-Fi Communication Module allows NeuLog sensor measurements to be performed through any device which uses Wi-Fi technology.
  • Intuitive and user friendly.
  • Allows saving and loading experiments.
  • Can be operated in client mode through the local router enabling also surfing to other sites.
  • No need for a USB module; it is part of Wi-Fi module.
  • Should be connected to a power supply or to a BAT-200 battery module.
  • The teacher can view student experiments in remote view.
  • Students can view the teacher's experiment on their table.
  • Factory-calibrated.
Specifications
Type Sensor
Requirements Power Source
For Use With (Application) Works with up to 5 sensors at once