Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NeuN Polyclonal Antibody

Rabbit Polyclonal Antibody
$553.50 - $1763.00
Specifications
| Antigen | NeuN |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5143507
|
Invitrogen™
PA5143507 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143605
![]() |
Invitrogen™
PA5143605 |
100 μL |
Each for $553.50
|
|
|||||
Description
NeuN Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.Specifications
| NeuN | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| RBFOX3 | |
| D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3 | |
| RBFOX3 | |
| Primary | |
| Affinity Chromatography |
| 1 mg/mL | |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| A6NFN3, Q8BIF2 | |
| 146713, 287847, 52897 | |
| Peptide corresponding to amino acids 5-24 of human FOX3 coupled to KLH. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title