Learn More
Invitrogen™ NeuN Polyclonal Antibody

Description
Positive Samples: Mouse brain, Rat brain Immunogen sequence: MAQPYPPAQY PPPPQNGIPA EYAPPPPHPT QDYSGQTPVP TEHGMTLYTP AQTHPEQPGS EASTQPIAGT QTVPQTDEAA QTDSQPLHPS DPTEKQQPKR LHVSNIPFRF RDPDLRQMFG QFGKILDVEI IFNERGSKGF GFVTFETSSD ADRAREKLNG TIVEGRKIEV NNATARVMTN KKTGNPYTNG WKLNPVVGAV YGPEFYAVTG FPYPTTGTAV.
Specifications
Specifications
| Antigen | NeuN |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.72 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RBFOX3 |
| Gene Accession No. | A6NFN3, Q8BIF2 |
| Gene Alias | D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3 |
| Gene Symbols | RBFOX3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.