Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (9.2.27), eBioscience™, Invitrogen™

Catalog No. 14650437
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-650-437 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-650-437 Supplier Invitrogen™ Supplier No. 14650437
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This 9.2.27 MAb reacts with human neural/glial antigen 2, which is also known as melanoma chondroitin sulfate proteoglycan (MCSP). This antigen is composed of a 250 kDa N-linked glycoprotein and a >450 kDa proteoglycan. NG2 is present on the surface of >90% of malignant melanomas in addition to some non-melanomic tumors. Moreover, NG2 is expressed on glioma cells, as well as on developing and adult oligodendrocyte precursor cells. Studies have demonstrated the involvement of this protein in tumor cell proliferation, adhesion, migration, and invasion. NG2 also functions as a coreceptor for alpha4beta1 integrin. Finally, expression of NG2 can be used as a prognostic marker for disorders such as acral letiginous melanoma and infantile acute myeloid leukemia. The 9.2.27 antibody has been reported to suppress melanoma tumor growth.

A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neural/Glial Antigen 2 (NG2)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9.2.27
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cspg4
Gene Accession No. Q6UVK1
Gene Alias 4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2
Gene Symbols Cspg4
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1464
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.