Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Neuraminidase (C. perfringens)

Catalog No. 89954019 Shop All Abnova Corporation Products
Click to view available options
:
5 Units

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Name Neuraminidase (C. perfringens)
Preparation Method Native protein chromatographically purified from Clostridium perfringens
Quantity 5 Units
Storage Requirements Store at 4°C on dry atmosphere.
Regulatory Status RUO
Common Name NEURAMINIDASE
Assays Neuraminidase Assay; Study of glycoproteins in protein chemistry and cell biology.
Biological Activity ≥10 units per mg protein. One Unit releases one micromole of sialic acid per minute at 37°C, pH 5.0, from bovine submaxillary mucin.
Species Clostridium species
Protein Tag None
Expression System Native protein chromatographically purified from Clostridium perfringens
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.