Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium perfringens Neuraminidase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA185712
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185712 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185712 Supplier Invitrogen™ Supplier No. PA185712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit anti Bacterial sialidase antibody recognizes sialidase, also known as neuraminidase, from bacterial species Clostridium perfringens.

Sialidase, also known as Neuraminidase, is a secreted protein from bacterial species Clostridium perfringens that catalyses hydrolysis of glycosidic linkages on sialic acid residues in many biomolecules. It is suggested to be involved in the pathogenesis of microbial infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium perfringens Neuraminidase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias Sialidase
Host Species Rabbit
Immunogen Native sialidase from Clostridium perfringens.
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.