Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuregulin-1/NRG1 Antibody (NRG1/2752), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279847G
Description
Neuregulin-1/NRG1 Monoclonal specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Neuregulin-1/NRG1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NRG1 | |
Recombinant fragment of human Neuregulin-1/NRG1 protein (around aa 21-242) (exact sequence is proprietary) (Uniprot: Q02297) | |
0.1 mL | |
Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
Human | |
IgG |
ELISA, Immunocytochemistry, Immunofluorescence | |
NRG1/2752 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction