Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuregulin-1 Rat anti-Human, Clone: 4F10, MilliporeSigma™

Catalog No. MABN229225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2292-25 25 μg
MABN2292-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2292-25 Supplier MilliporeSigma Supplier No. MABN229225UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Neuregulin-1 clone 4F10 is a highly specific rat monoclonal antibody that targets Typ III NRG1 beta-1 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuregulin-1
Applications Western Blot
Classification Monoclonal
Clone 4F10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols NRG1,GGF,HGL,HRGA,NDF,SMDF
Host Species Rat
Immunogen A linear peptide corresponding to 10 amino acids from the N-terminal, extracellular domain of human Neuregulin 1, isoform 6.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_039250
Test Specificity Clone 4F10 is a rat monoclonal antibody that detects human Neuregulin-1. It targets an epitope within 10 amino acids from the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.