Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurensin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543451
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543451 Supplier Invitrogen™ Supplier No. PA543451
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LLLLLGVAAL TTGYAVPPKL EGIGEGEFLV LDQRAADYNQ ALGTCRLAGT Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 83%; Horse: 93%; Human: 100%; Pig: 92%; Rabbit: 100%.

Neurensin-2 (NRSN2) is a 204 amino acid multi-pass membrane protein belonging to the VMP family that may be involved in the transport and maintenance of vesicles. Expressed in brain, Neurensin-2 localizes to cell bodies of hippocampus, diagonal band, amygdaloid nucleus, and habenula nucleus, and is a potential tumor suppressor gene and candidate biomarker for long-term survival in patients with hepatocellular carcinoma (HCC). The gene encoding Neurensin-2 maps to human chromosome 20, which comprises approximately 2% of the human genome, contains nearly 63 million bases and encodes over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought important for seminal production and may be potential targets for male contraception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurensin 2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NRSN2
Gene Accession No. Q9GZP1
Gene Alias C20orf98; dJ1103G7.6; FLJ23329; Gm123; neurensin 2; Neurensin2; Neurensin-2; NRSN2
Gene Symbols NRSN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human NRSN2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80023
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.