Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurexin 2 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11104R

Description
Neuronal cell surface protein that may be involved in cell recognition and cell adhesion.Specifications
Neurexin 2 | |
Polyclonal | |
Unconjugated | |
NRXN2 | |
NRXN2 | |
KLH conjugated synthetic peptide derived from human Neurexin 2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P58401, Q63374, Q9P2S2 | |
Rabbit | |
Protein A | |
RUO | |
116595, 18190, 9379 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction